Tisk…

Fmoc-N-amido-PEG10-Acid

Dodavatel: Apollo Scientific

Synonyms: Fmoc-NH-PEG10-CH2CH2COOH

BIPG1404-100MG
APOSBIPG1404-100MGEA 622 CZK
APOSBIPG1404-100MG
Fmoc-N-amido-PEG10-Acid
Fmoc-N-amido-PEG10-Acid
Vzorec: C₃₈H₅₇NO₁₄
MW: 751,857 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28122954
CAS číslo: 2101563-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR