Cbz-N-amido-PEG2-acid
Dodavatel: Apollo Scientific
Synonyms:
Cbz-NH-PEG2-C2-acid, Cbz-n-amido-peg2-acid
BIPG1361-1G
BIPG1361-250MG
APOSBIPG1361-1GEA
5920
CZK
APOSBIPG1361-1G
APOSBIPG1361-250MG
Cbz-N-amido-PEG2-acid
Cbz-N-amido-PEG2-acid
Vzorec:
C₁₅H₂₁NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD28142483 CAS číslo: 1347750-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...