Biotin-PEG6-Acid
Dodavatel: Apollo Scientific
|
Varování
|
BIPG1200-100MG
BIPG1200-250MG
APOSBIPG1200-100MGEA
6620
CZK
APOSBIPG1200-100MG
APOSBIPG1200-250MG
Biotin-PEG6-Acid
Biotin-PEG6-Acid
Vzorec:
C₂₅H₄₅N₃O₁₀S MW: 579.704 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16875661 CAS číslo: 1352814-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...