Biotin-PEG2-Acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(2-(2-(5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)ethoxy)ethoxy)propanoic acid
BIPG1197-100MG
BIPG1197-250MG
APOSBIPG1197-100MGEA
6020
CZK
APOSBIPG1197-100MG
APOSBIPG1197-250MG
Biotin-PEG2-Acid
Biotin-PEG2-Acid
Vzorec:
C₁₇H₂₉N₃O₆S MW: 403,494 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22056305 CAS číslo: 1365655-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...