Tisk…

Azido-PEG6-Acid

Dodavatel: Apollo Scientific
Varování

BIPG1108-100MG BIPG1108-50MG BIPG1108-250MG
APOSBIPG1108-100MGEA 1950 CZK
APOSBIPG1108-100MG APOSBIPG1108-50MG APOSBIPG1108-250MG
Azido-PEG6-Acid
Azido-PEG6-Acid
Vzorec: C₁₅H₂₉N₃O₈
MW: 379,406 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22056306
CAS číslo: 361189-66-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR