Amino-PEG4-CH2CO2H
Dodavatel: Apollo Scientific
BIPG1064-250MG
BIPG1064-100MG
APOSBIPG1064-250MGEA
3480
CZK
APOSBIPG1064-250MG
APOSBIPG1064-100MG
Amino-PEG4-CH2CO2H
Amino-PEG4-CH2CO2H
Vzorec:
C₁₀H₂₁NO₆ MW: 251,277 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27977508 CAS číslo: 95071-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...