Amino-PEG11-Amine
Dodavatel: Apollo Scientific
Synonyms:
3,6,9,12,15,18,21,24,27,30,33-Undecaoxapentatriacontane-1,35-diamine
BIPG1062-250MG
BIPG1062-100MG
APOSBIPG1062-250MGEA
4300
CZK
APOSBIPG1062-250MG
APOSBIPG1062-100MG
Amino-PEG11-Amine
Amino-PEG11-Amine
Vzorec:
C₂₄H₅₂N₂O₁₁ MW: 544,676 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28122961 CAS číslo: 479200-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...