Tisk…

Amino-PEG24-alcohol

Dodavatel: Apollo Scientific
Varování

BIPG1053-100MG BIPG1053-50MG
APOSBIPG1053-100MGEA 14880 CZK
APOSBIPG1053-100MG APOSBIPG1053-50MG
Amino-PEG24-alcohol
Amino-PEG24-alcohol
Vzorec: C₄₈H₉₉NO₂₄
MW: 1074,29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD21363266
CAS číslo: 933789-97-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR