4-Fluoro-3-nitrophenyl azide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
BIPA100-100MG
BIPA100-1G
APOSBIPA100-100MGEA
3940
CZK
APOSBIPA100-100MG
APOSBIPA100-1G
4-Fluoro-3-nitrophenyl azide
4-Fluoro-3-nitrophenyl azide
Photo affinity labelling
Vzorec:
C₆H₃FN₄O₂ MW: 182.112 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00036215 CAS číslo: 28166-06-5 EINECS: 248-878-6 UN: 2926 ADR: 4.1+6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...