Tisk…

5-Bromo-4-chloro-3-indolyl-N-acetaminide

Dodavatel: Apollo Scientific
Varování

Synonyms: X-acetaminide

BIMB1683-1G BIMB1683-2.5G BIMB1683-500MG
APOSBIMB1683-1GEA 4960 CZK
APOSBIMB1683-1G APOSBIMB1683-2.5G APOSBIMB1683-500MG
5-Bromo-4-chloro-3-indolyl-N-acetaminide
5-Bromo-4-chloro-3-indolyl-N-acetaminide
Vzorec: C₁₀H₈BrClN₂O
MW: 287,54 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14155891
CAS číslo: 1330750-19-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR