6-Chloro-3-indolyl-α-D-galactopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Salmon-α-Gal
BIMB1326-100MG
BIMB1326-250MG
APOSBIMB1326-100MGEA
5960
CZK
APOSBIMB1326-100MG
APOSBIMB1326-250MG
6-Chloro-3-indolyl-α-D-galactopyranoside
6-Chloro-3-indolyl-α-D-galactopyranoside
Vzorec:
C₁₄H₁₆ClNO₆ MW: 329,733 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02683908 CAS číslo: 198402-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...