Tisk…

5-Bromo-4-chloro-3-indolyl-α-D-glucopyranoside

Dodavatel: Apollo Scientific
Varování

Synonyms: X-α-Glucoside

BIMB1190-100MG BIMB1190-250MG
APOSBIMB1190-100MGEA 2900 CZK
APOSBIMB1190-100MG APOSBIMB1190-250MG
5-Bromo-4-chloro-3-indolyl-α-D-glucopyranoside
5-Bromo-4-chloro-3-indolyl-α-D-glucopyranoside
Vzorec: C₁₄H₁₅BrClNO₆
MW: 408,629 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00155606
CAS číslo: 108789-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR