Methyl 6-amino-6-deoxy-a-D-glucopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
BIM6003-100MG
BIM6003-250MG
BIM6003-500MG
APOSBIM6003-100MGEA
4140
CZK
APOSBIM6003-100MG
APOSBIM6003-250MG
APOSBIM6003-500MG
Methyl 6-amino-6-deoxy-a-D-glucopyranoside
Methyl 6-amino-6-deoxy-a-D-glucopyranoside
Vzorec:
C₇H₁₅NO₅ MW: 193,198 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038151 CAS číslo: 5155-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...