Bis(4-methylumbelliferyl) phosphate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
bis-MUP
BIM2048-1G
BIM2048-250MG
APOSBIM2048-1GEA
10750
CZK
APOSBIM2048-1G
APOSBIM2048-250MG
Bis(4-methylumbelliferyl) phosphate
Bis(4-methylumbelliferyl) phosphate
Vzorec:
C₂₀H₁₅O₈P MW: 414.302 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00076128 CAS číslo: 51379-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...