4-Methylumbelliferyl nonanoate
Dodavatel: Apollo Scientific
BIM2046-1G
BIM2046-250MG
APOSBIM2046-1GEA
4140
CZK
APOSBIM2046-1G
APOSBIM2046-250MG
4-Methylumbelliferyl nonanoate
4-Methylumbelliferyl nonanoate
Vzorec:
C₁₉H₂₄O₄ MW: 316,391 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00051220 CAS číslo: 18319-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...