Tisk…

4-Methylumbelliferyl palmitate

Dodavatel: Apollo Scientific
Varování

BIM1102-100MG BIM1102-500MG BIM1102-250MG
APOSBIM1102-100MGEA 1580 CZK
APOSBIM1102-100MG APOSBIM1102-500MG APOSBIM1102-250MG
4-Methylumbelliferyl palmitate
4-Methylumbelliferyl palmitate
Vzorec: C₂₆H₃₈O₄
MW: 414.578 g/mol
Bod tání: 77 °C
Storage Temperature: Freezer
MDL Number: MFCD00037612
CAS číslo: 17695-48-6
EINECS: 241-695-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR