Tisk…

3-Isothiocyanophenylboronic acid

Dodavatel: Apollo Scientific
Nebezpečí

BII109-10MG BII109-25MG
APOSBII109-10MGEA 10350 CZK
APOSBII109-10MG APOSBII109-25MG
3-Isothiocyanophenylboronic acid
3-Isothiocyanophenylboronic acid

Amine reactive -Iodinatable

Vzorec: C₇H₆BNO₂S
Bod tání: 166…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00467363
CAS číslo: 133887-74-8
UN: 2928
ADR: 6.1+8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR