Tisk…

D-Galactosamine pentaacetate

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,2,3,4,6-Penta-O-acetyl-D-galactosamine, (3R,4R,5R,6R)-3-Acetamido-6-(acetoxymethyl)tetrahydro-2H-pyran-2,4,5-triyl triacetate

BIG5003-1G BIG5003-5G
APOSBIG5003-1GEA 622 CZK
APOSBIG5003-1G APOSBIG5003-5G
D-Galactosamine pentaacetate
D-Galactosamine pentaacetate
Vzorec: C₁₆H₂₃NO₁₀
MW: 389,355 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00050926
CAS číslo: 76375-60-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR