Tisk…

L-Glucono-1,5-lactone ≥85%

Dodavatel: Apollo Scientific

BIG1121-100MG BIG1121-1G BIG1121-250MG
APOSBIG1121-100MGEA 6850 CZK
APOSBIG1121-100MG APOSBIG1121-1G APOSBIG1121-250MG
L-Glucono-1,5-lactone ≥85%
L-Glucono-1,5-lactone
Vzorec: C₆H₁₀O₆
Bod tání: 135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067388
CAS číslo: 52153-09-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR