Dovitinib lactate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
CHIR-258, TKI-258, 4-Amino-5-fluoro-3-(6-(4-methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)quinolin-2(1H)-one 2-hydroxypropanoate
BIFK0016-5MG
APOSBIFK0016-5MGEA
1990
CZK
APOSBIFK0016-5MG
Dovitinib lactate
Dovitinib lactate
Lactate Salt, >99%
Vzorec:
C₂₁H₂₁FN₆O·C₃H₆O₃ MW: 482,507 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16038905 CAS číslo: 692737-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...