Tisk…

3-Fluoro-DL-phenylalanine

Dodavatel: Apollo Scientific

BIF4028-1G BIF4028-5G
APOSBIF4028-1GEA 622 CZK
APOSBIF4028-1G APOSBIF4028-5G
3-Fluoro-DL-phenylalanine
3-Fluoro-DL-phenylalanine
Vzorec: C₉H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD00004273
CAS číslo: 456-88-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR