Tisk…

5-Bromo-4-chloro-3-indolyl α-D-N-acetylneuraminic acid sodium salt

Dodavatel: Apollo Scientific
Varování

BICS0105-10MG BICS0105-25MG BICS0105-50MG
APOSBICS0105-10MGEA 4140 CZK
APOSBICS0105-10MG APOSBICS0105-25MG APOSBICS0105-50MG
5-Bromo-4-chloro-3-indolyl α-D-N-acetylneuraminic acid sodium salt
5-Bromo-4-chloro-3-indolyl α-D-N-acetylneuraminic acid sodium salt
Vzorec: C₁₉H₂₁BrClN₂O₉·Na
Bod tání: >183 °C
Storage Temperature: Freezer
MDL Number: MFCD00792680
CAS číslo: 160369-85-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR