Tisk…

4-Br debranone

Dodavatel: Apollo Scientific

Synonyms: N,4-Br debranone

BICR144-10MG BICR144-50MG
APOSBICR144-10MGEA 9860 CZK
APOSBICR144-10MG APOSBICR144-50MG
4-Br debranone
4-Br debranone
Vzorec: C₁₁H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD28556859
CAS číslo: 1332863-94-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR