Tisk…

(S)-Boc-α-methyl-phenylglycine

Dodavatel: Apollo Scientific
Varování

Synonyms: N,(S)-Boc-α-methyl-phenylglycine

BICR126-1G BICR126-5G
APOSBICR126-1GEA 12630 CZK
APOSBICR126-1G APOSBICR126-5G
(S)-Boc-α-methyl-phenylglycine
(S)-Boc-α-methyl-phenylglycine
Vzorec: C₁₄H₁₉NO₄
Storage Temperature: Freezer
MDL Number: MFCD11052916
CAS číslo: 802541-88-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR