Tisk…

(S)-α-Methyl-phenylglycine

Dodavatel: Apollo Scientific
Varování

Synonyms: N,(S)-α-methyl-phenylglycine

BICR107-1G BICR107-5G
APOSBICR107-1GEA 11080 CZK
APOSBICR107-1G APOSBICR107-5G
(S)-α-Methyl-phenylglycine
(S)-α-Methyl-phenylglycine
Vzorec: C₉H₁₁NO₂
MW: 165,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371947
CAS číslo: 13398-26-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR