(3R,4R)-3,4-dihydroxycyclohexa-1,5-diene-1-carboxylic acid
Dodavatel: Apollo Scientific
BICR104-1G
BICR104-500MG
APOSBICR104-1GEA
30780
CZK
APOSBICR104-1G
APOSBICR104-500MG
(3R,4R)-3,4-dihydroxycyclohexa-1,5-diene-1-carboxylic acid
(3R,4R)-3,4-dihydroxycyclohexa-1,5-diene-1-carboxylic acid
Vzorec:
C₇H₈O₄ Storage Temperature: Ambient |
MDL Number:
MFCD06797325 CAS číslo: 108646-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...