Tisk…

D-Homopropargylglycine

Dodavatel: Apollo Scientific
Varování

Synonyms: N,D-Homopropargylglycine

BICR101-200MG BICR101-500MG BICR101-100MG
APOSBICR101-200MGEA 13030 CZK
APOSBICR101-200MG APOSBICR101-500MG APOSBICR101-100MG
D-Homopropargylglycine
D-Homopropargylglycine
Vzorec: C₆H₉NO₂
MW: 127,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052900
CAS číslo: 211054-02-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR