Tisk…

2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl trichloroacetimidate ≥95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,3,4,6-Tetra-O-acetyl-1-O-trichloroacetimidoyl-α-D-mannopyranoside, (2R,3R,4S,5S,6R)-2-(Acetoxymethyl)-6-(2,2,2-trichloro-1-iminoethoxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate

BICL4375-1G BICL4375-2G BICL4375-500MG
APOSBICL4375-1GEA 21890 CZK
APOSBICL4375-1G APOSBICL4375-2G APOSBICL4375-500MG
2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl trichloroacetimidate ≥95%
2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl trichloroacetimidate
Vzorec: C₁₆H₂₀Cl₃NO₁₀
Storage Temperature: Freezer
MDL Number: MFCD04039685
CAS číslo: 121238-27-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR