Tisk…

4-Isothiocyanatophenyl α-D-mannopyranoside ≥97%

Dodavatel: Apollo Scientific

Synonyms: (2R,3S,4S,5S,6R)-2-(Hydroxymethyl)-6-(4-isothiocyanatophenoxy)tetrahydro-2H-pyran-3,4,5-triol, α-D-Mannopyranosyl 4-phenylisothiocyanate

BICL4346-25MG BICL4346-50MG BICL4346-100MG
APOSBICL4346-25MGEA 7210 CZK
APOSBICL4346-25MG APOSBICL4346-50MG APOSBICL4346-100MG
4-Isothiocyanatophenyl α-D-mannopyranoside ≥97%
4-Isothiocyanatophenyl α-D-mannopyranoside
Vzorec: C₁₃H₁₅NO₆S
MW: 313,326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070374
CAS číslo: 96345-79-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR