2-(Dihydrogen phosphate) D-glucose ≥95%
Dodavatel: Apollo Scientific
Synonyms:
D-Glucose 2-phosphate
BICL4330-1MG
BICL4330-2MG
BICL4330-5MG
APOSBICL4330-1MGEA
14880
CZK
APOSBICL4330-1MG
APOSBICL4330-2MG
APOSBICL4330-5MG
2-(Dihydrogen phosphate) D-glucose ≥95%
2-(Dihydrogen phosphate) D-glucose
Vzorec:
C₆H₁₃O₉P MW: 260,136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28397795 CAS číslo: 67101-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...