Tisk…

Dihydroartemisinin O-β-D-glucuronide ≥95%

Dodavatel: Apollo Scientific

BICL4328-1MG BICL4328-2MG
APOSBICL4328-1MGEA 12370 CZK
APOSBICL4328-1MG APOSBICL4328-2MG
Dihydroartemisinin O-β-D-glucuronide ≥95%
Dihydroartemisinin O-β-D-glucuronide
Storage Temperature: Ambient
MDL Number: MFCD08063733
CAS číslo: 198976-06-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR