1,6:3,4-Dianhydro-2-O-benzyl-β-D-altropyranose ≥98%
Dodavatel: Apollo Scientific
Synonyms:
Altro-erny epoxide 2-O-benzyl-
BICL4327-500MG
BICL4327-100MG
BICL4327-250MG
APOSBICL4327-500MGEA
38350
CZK
APOSBICL4327-500MG
APOSBICL4327-100MG
APOSBICL4327-250MG
1,6:3,4-Dianhydro-2-O-benzyl-β-D-altropyranose ≥98%
1,6:3,4-Dianhydro-2-O-benzyl-β-D-altropyranose
Vzorec:
C₁₃H₁₄O₄ Storage Temperature: Ambient |
MDL Number:
MFCD15144986 CAS číslo: 213594-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...