2,5-Anhydro-L-iditol ≥98%
Dodavatel: Apollo Scientific
BICL4321-10MG
BICL4321-25MG
BICL4321-5MG
APOSBICL4321-10MGEA
16530
CZK
APOSBICL4321-10MG
APOSBICL4321-25MG
APOSBICL4321-5MG
2,5-Anhydro-L-iditol ≥98%
2,5-Anhydro-L-iditol
Vzorec:
C₆H₁₂O₅ MW: 164,156 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09702436 CAS číslo: 28218-55-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...