6-Amino-6-deoxy-D-glucopyranose, hydrochloride ≥98%
Dodavatel: Apollo Scientific
BICL4320-100MG
BICL4320-25MG
BICL4320-50MG
APOSBICL4320-100MGEA
34390
CZK
APOSBICL4320-100MG
APOSBICL4320-25MG
APOSBICL4320-50MG
6-Amino-6-deoxy-D-glucopyranose, hydrochloride ≥98%
6-Amino-6-deoxy-D-glucopyranose, hydrochloride
Vzorec:
C₆H₁₃NO₅·ClH Storage Temperature: Ambient |
MDL Number:
MFCD00051996 CAS číslo: 4460-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...