2-O-(Carboxymethyl)-D-glucose ≥95%
Dodavatel: Apollo Scientific
BICL4306-5MG
BICL4306-1MG
BICL4306-2MG
APOSBICL4306-5MGEA
30220
CZK
APOSBICL4306-5MG
APOSBICL4306-1MG
APOSBICL4306-2MG
2-O-(Carboxymethyl)-D-glucose ≥95%
2-O-(Carboxymethyl)-D-glucose
Storage Temperature:
Ambient |
CAS číslo:
95350-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...