6-O-(α-D-Galactopyranosyl)-D-galactopyranose ≥95%
Dodavatel: Apollo Scientific
BICL4303-10MG
BICL4303-25MG
BICL4303-50MG
APOSBICL4303-10MGEA
12370
CZK
APOSBICL4303-10MG
APOSBICL4303-25MG
APOSBICL4303-50MG
6-O-(α-D-Galactopyranosyl)-D-galactopyranose ≥95%
6-O-(α-D-Galactopyranosyl)-D-galactopyranose
Storage Temperature:
Ambient |
CAS číslo:
902-54-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...