Tolmetin acyl-β-D-glucuronide ≥95%
Dodavatel: Apollo Scientific
BICL4301-1MG
BICL4301-2MG
BICL4301-5MG
APOSBICL4301-1MGEA
9560
CZK
APOSBICL4301-1MG
APOSBICL4301-2MG
APOSBICL4301-5MG
Tolmetin acyl-β-D-glucuronide ≥95%
Tolmetin acyl-β-D-glucuronide
Storage Temperature:
Ambient |
CAS číslo:
71595-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...