Tisk…
4-O-α-D-Galactopyranosyl-D-sucrose ≥95%
Dodavatel: Apollo Scientific
Synonyms: Nigellamose, β-D-Fructofuranosyl 4-O-α-D-galactopyranosyl-α-D-glucopyranoside
BICL4294-5MG BICL4294-1MG BICL4294-2MG
APOSBICL4294-5MGEA 22290 CZK
APOSBICL4294-5MG APOSBICL4294-1MG APOSBICL4294-2MG
4-O-α-D-Galactopyranosyl-D-sucrose ≥95%
4-O-α-D-Galactopyranosyl-D-sucrose
Storage Temperature: Ambient
CAS číslo: 155154-22-6