Tisk…

Octanoyl D-glucopyranoside ≥98%

Dodavatel: Apollo Scientific

BICL4286-1G BICL4286-2G
APOSBICL4286-1GEA 11970 CZK
APOSBICL4286-1G APOSBICL4286-2G
Octanoyl D-glucopyranoside ≥98%
Octanoyl D-glucopyranoside
Vzorec: C₁₄H₂₆O₇
Storage Temperature: Ambient
MDL Number: MFCD28122839
CAS číslo: 60415-65-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR