Tisk…

4-Methoxyphenyl 6-O-tert-butyldiphenylsilyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98%

Dodavatel: Apollo Scientific

Synonyms: p-Methoxyphenyl 6-O-TBDPS-3,4-O-isopropylidene-β-D-galactopyranoside

BICL4284-1G BICL4284-250MG BICL4284-500MG
APOSBICL4284-1GEA 36040 CZK
APOSBICL4284-1G APOSBICL4284-250MG APOSBICL4284-500MG
4-Methoxyphenyl 6-O-tert-butyldiphenylsilyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98%
4-Methoxyphenyl 6-O-tert-butyldiphenylsilyl-3,4-O-isopropylidene-β-D-galactopyranoside
Vzorec: C₃₂H₄₀O₇Si
MW: 564,741 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28122837
CAS číslo: 503303-07-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR