Methyl 2-deoxy-2-fluoro-L-arabinofuranoside ≥95%
Dodavatel: Apollo Scientific
|
Varování
|
BICL4282-100MG
BICL4282-250MG
BICL4282-50MG
APOSBICL4282-100MGEA
15150
CZK
APOSBICL4282-100MG
APOSBICL4282-250MG
APOSBICL4282-50MG
Methyl 2-deoxy-2-fluoro-L-arabinofuranoside ≥95%
Methyl 2-deoxy-2-fluoro-L-arabinofuranoside
Vzorec:
C₆H₁₁FO₄ MW: 166,148 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09833554 CAS číslo: 442514-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...