Tisk…

1,3,4,6-Tetra-O-acetyl-2-deoxy-2-iodo-β-D-galactopyranose ≥98%

Dodavatel: Apollo Scientific

BICL4275-100MG BICL4275-250MG
APOSBICL4275-100MGEA 14350 CZK
APOSBICL4275-100MG APOSBICL4275-250MG
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-iodo-β-D-galactopyranose ≥98%
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-iodo-β-D-galactopyranose
Vzorec: C₁₄H₁₉IO₉
MW: 458,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28140554
CAS číslo: 141510-66-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR