Tisk…

Hesperetin 3'-O-β-D-glucuronide ≥95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(3,4-Dihydro-5,7-dihydroxy-4-oxo-2H-1-benzopyran-2-yl)-2-methoxyphenyl β-D-glucopyranosiduronic acid

BICL4274-10MG BICL4274-5MG BICL4274-1MG
APOSBICL4274-10MGEA 13960 CZK
APOSBICL4274-10MG APOSBICL4274-5MG APOSBICL4274-1MG
Hesperetin 3'-O-β-D-glucuronide ≥95%
Hesperetin 3'-O-β-D-glucuronide
Vzorec: C₂₂H₂₂O₁₂
Storage Temperature: Freezer
MDL Number: MFCD28122831
CAS číslo: 1237479-05-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR