Tisk…

Caffeic acid 3-O-β-D-glucuronide ≥98%

Dodavatel: Apollo Scientific
Varování

BICL4272-2MG BICL4272-5MG BICL4272-1MG
APOSBICL4272-2MGEA 12730 CZK
APOSBICL4272-2MG APOSBICL4272-5MG APOSBICL4272-1MG
Caffeic acid 3-O-β-D-glucuronide ≥98%
Caffeic acid 3-O-β-D-glucuronide
Vzorec: C₁₅H₁₆O₁₀
Storage Temperature: Freezer
MDL Number: MFCD28122829
CAS číslo: 1093679-73-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR