α-D-Xylopyranosyl azide ≥98%
Dodavatel: Apollo Scientific
BICL4270-100MG
BICL4270-25MG
BICL4270-50MG
APOSBICL4270-100MGEA
36040
CZK
APOSBICL4270-100MG
APOSBICL4270-25MG
APOSBICL4270-50MG
α-D-Xylopyranosyl azide ≥98%
α-D-Xylopyranosyl azide
Vzorec:
C₅H₉N₃O₄ MW: 175,143 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28122827 CAS číslo: 100842-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...