Tisk…

4-O-β-D-Galactopyranosyl-D-sucrose ≥97%

Dodavatel: Apollo Scientific

Synonyms: α-D-Lactopyranosyl-β-D-fructofuranoside, β-D-Fructofuranosyl 4-O-β-D-galactopyranosyl-α-D-glucopyranoside

BICL4263-5MG BICL4263-1MG BICL4263-2MG
APOSBICL4263-5MGEA 24270 CZK
APOSBICL4263-5MG APOSBICL4263-1MG APOSBICL4263-2MG
4-O-β-D-Galactopyranosyl-D-sucrose ≥97%
4-O-β-D-Galactopyranosyl-D-sucrose
Vzorec: C₁₈H₃₂O₁₆
MW: 504,437 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272612
CAS číslo: 87419-56-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR