Tisk…

Diethyl dithioacetal D-galactose ≥98%

Dodavatel: Apollo Scientific

BICL4251-1G BICL4251-2G
APOSBICL4251-1GEA 14350 CZK
APOSBICL4251-1G APOSBICL4251-2G
Diethyl dithioacetal D-galactose ≥98%
Diethyl dithioacetal D-galactose
Vzorec: C₁₀H₂₂O₅S₂
Storage Temperature: Ambient
MDL Number: MFCD00069630
CAS číslo: 5463-33-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR