Tisk…

3-Nitrophenyl α-D-galactopyranoside ≥98%

Dodavatel: Apollo Scientific

Synonyms: (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(3-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

BICL4247-100MG BICL4247-250MG
APOSBICL4247-100MGEA 3060 CZK
APOSBICL4247-100MG APOSBICL4247-250MG
3-Nitrophenyl α-D-galactopyranoside ≥98%
3-Nitrophenyl α-D-galactopyranoside
Vzorec: C₁₂H₁₅NO₈
MW: 301,249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067358
CAS číslo: 52571-71-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR