Benzyl 2-acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-α-D-galactopyranoside ≥98%
Dodavatel: Apollo Scientific
Synonyms:
β-D-Gal-(1→3)-α-D-GalNAc-1→OCH2Ph, Benzyl 2-(acetylamino)-2-deoxy-3-O-(β-D-galactopyranosyl)-α-D-galactopyranoside, Benzyl 3-O-(β-D-galactopyranosyl)-α-D-GalNAc
BICL4241-10MG
BICL4241-25MG
BICL4241-50MG
APOSBICL4241-10MGEA
11970
CZK
APOSBICL4241-10MG
APOSBICL4241-25MG
APOSBICL4241-50MG
Benzyl 2-acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-α-D-galactopyranoside ≥98%
Benzyl 2-acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-α-D-galactopyranoside
Vzorec:
C₂₁H₃₁NO₁₁ MW: 473,471 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057916 CAS číslo: 3554-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...