Tisk…

3-O-Benzyl-1,2:5,6-di-O-isopropylidene-α-D-allofuranose ≥98%

Dodavatel: Apollo Scientific

Synonyms: (3aR,5R,6R,6aR)-6-(Benzyloxy)-5-((R)-2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxole

BICL4234-25G
APOSBICL4234-25GEA 0 CZK
APOSBICL4234-25G
3-O-Benzyl-1,2:5,6-di-O-isopropylidene-α-D-allofuranose ≥98%
3-O-Benzyl-1,2:5,6-di-O-isopropylidene-α-D-allofuranose
Vzorec: C₁₉H₂₆O₆
MW: 350,406 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704028
CAS číslo: 22331-21-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR